4-Bromo-2,5-difluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27924-250MG
APOSPC27924-250MGEA 79.9 EUR
APOSPC27924-250MG
4-Bromo-2,5-difluorobenzyl fluoride 95%
4-Bromo-2,5-difluorobenzyl fluoride
Formel: C₇H₄BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31664905
CAS-Nummer: 2244083-92-7
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR