2-Bromo-4-fluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27927-250MG
APOSPC27927-250MGEA 79.9 EUR
APOSPC27927-250MG
2-Bromo-4-fluorobenzyl fluoride 95%
2-Bromo-4-fluorobenzyl fluoride
Formel: C₇H₅BrF₂
Molecular Weight: 207.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22491732
CAS-Nummer: 1823057-15-3
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR