4-Bromo-2,6-difluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27936-250MG
APOSPC27936-250MGEA 79.9 EUR
APOSPC27936-250MG
4-Bromo-2,6-difluorobenzyl fluoride 95%
4-Bromo-2,6-difluorobenzyl fluoride
Formel: C₇H₄BrF₃
Molecular Weight: 225.006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664910
CAS-Nummer: 2244087-29-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR