2-Hydrazinyl-3,5-bis(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC27940-1G
APOSPC27940-1GEA 1210 EUR
APOSPC27940-1G
2-Hydrazinyl-3,5-bis(trifluoromethyl)pyridine 95%
2-Hydrazinyl-3,5-bis(trifluoromethyl)pyridine
Formel: C₇H₅F₆N₃
Molecular Weight: 245,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068122
CAS-Nummer: 129015-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR