2-Hydrazinyl-4,6-bis(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
PC27941-1G
APOSPC27941-1GEA
1210
EUR
APOSPC27941-1G
2-Hydrazinyl-4,6-bis(trifluoromethyl)pyridine 95%
2-Hydrazinyl-4,6-bis(trifluoromethyl)pyridine
Formel:
C₇H₅F₆N₃ Molecular Weight: 245,125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215672 CAS-Nummer: 176762-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...