(3E)-1-(4-Fluorophenyl)-3-(methoxyimino)propan-1-one 95%

Lieferant: Apollo Scientific

PC27942-1G
APOSPC27942-1GEA 1570 EUR
APOSPC27942-1G
(3E)-1-(4-Fluorophenyl)-3-(methoxyimino)propan-1-one 95%
(3E)-1-(4-Fluorophenyl)-3-(methoxyimino)propan-1-one
Formel: C₁₀H₁₀FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00127060
CAS-Nummer: 241488-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR