3-Chloro-2-(1H-pyrazol-4-yl)-5-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC27946-1G
APOSPC27946-1GEA 1570 EUR
APOSPC27946-1G
3-Chloro-2-(1H-pyrazol-4-yl)-5-(trifluoromethyl)pyridine 95%
3-Chloro-2-(1H-pyrazol-4-yl)-5-(trifluoromethyl)pyridine
Formel: C₉H₅ClF₃N₃
Molecular Weight: 247,604 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231891
CAS-Nummer: 246022-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR