(2E)-3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
Lieferant: Apollo Scientific
PC27947-1G
APOSPC27947-1GEA
1570
EUR
APOSPC27947-1G
(2E)-3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
(2E)-3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile
Formel:
C₁₁H₇ClF₃N₃O Molecular Weight: 289,641 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11656719 CAS-Nummer: 337919-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...