(2Z)-2-[(1E)-(Methoxyimino)methyl]-3-[4-(trifluoromethyl)phenyl]prop-2-enenitrile, tech

Lieferant: Apollo Scientific

PC27948-1G
APOSPC27948-1GEA 1570 EUR
APOSPC27948-1G
(2Z)-2-[(1E)-(Methoxyimino)methyl]-3-[4-(trifluoromethyl)phenyl]prop-2-enenitrile, tech
(2Z)-2-[(1E)-(Methoxyimino)methyl]-3-[4-(trifluoromethyl)phenyl]prop-2-enenitrile
Formel: C₁₂H₉F₃N₂O
Molecular Weight: 254,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663671
CAS-Nummer: 337919-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR