6-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
Lieferant: Apollo Scientific
PC27949-1G
APOSPC27949-1GEA
284
EUR
APOSPC27949-1G
6-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
6-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Formel:
C₁₂H₇FN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202736 CAS-Nummer: 31755-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...