1-Butyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

PC27952-1G
APOSPC27952-1GEA 1570 EUR
APOSPC27952-1G
1-Butyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech
1-Butyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one
Formel: C₁₆H₁₈FNO₂
Molecular Weight: 275,318 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871391
CAS-Nummer: 477846-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR