3-[Benzyl(methyl)amino]-1,1,1-trifluoropropan-2-ol 95%
Lieferant: Apollo Scientific
PC27953-1G
APOSPC27953-1GEA
1570
EUR
APOSPC27953-1G
3-[Benzyl(methyl)amino]-1,1,1-trifluoropropan-2-ol 95%
3-[Benzyl(methyl)amino]-1,1,1-trifluoropropan-2-ol
Formel:
C₁₁H₁₄F₃NO Molecular Weight: 233,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02186944 CAS-Nummer: 477847-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...