3-{[3-(Trifluoromethyl)phenyl]methyl}pentane-2,4-dione, tech

Lieferant: Apollo Scientific

PC27954-1G
APOSPC27954-1GEA 1570 EUR
APOSPC27954-1G
3-{[3-(Trifluoromethyl)phenyl]methyl}pentane-2,4-dione, tech
3-{[3-(Trifluoromethyl)phenyl]methyl}pentane-2,4-dione
Formel: C₁₃H₁₃F₃O₂
Molecular Weight: 258,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102560
CAS-Nummer: 691406-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR