(2E)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[3.2.2]nonan-4-one, tech

Lieferant: Apollo Scientific

PC27957-1G
APOSPC27957-1GEA 1570 EUR
APOSPC27957-1G
(2E)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[3.2.2]nonan-4-one, tech
(2E)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[3.2.2]nonan-4-one
Formel: C₁₅H₁₆FNO
Molecular Weight: 245,292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02081951
CAS-Nummer: 477848-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR