(2Z)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech

Lieferant: Apollo Scientific

PC27958-1G
APOSPC27958-1GEA 1570 EUR
APOSPC27958-1G
(2Z)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech
(2Z)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol
Formel: C₁₄H₁₆FNO
Molecular Weight: 233,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02081952
CAS-Nummer: 477848-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR