(2Z)-2-{[4-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech

Lieferant: Apollo Scientific

PC27959-1G
APOSPC27959-1GEA 1570 EUR
APOSPC27959-1G
(2Z)-2-{[4-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech
(2Z)-2-{[4-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol
Formel: C₁₅H₁₆F₃NO
Molecular Weight: 283,289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082072
CAS-Nummer: 477848-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR