(2Z)-2-[(2-Chloro-6-fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Lieferant: Apollo Scientific

PC27960-1G
APOSPC27960-1GEA 1570 EUR
APOSPC27960-1G
(2Z)-2-[(2-Chloro-6-fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(2-Chloro-6-fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₄H₁₃ClFNO
Molecular Weight: 265,711 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00109575
CAS-Nummer: 477848-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR