1-(4-Fluorophenyl)-4-(4-methylpiperazin-1-yl)butan-1-one, tech

Lieferant: Apollo Scientific

PC27961-1G
APOSPC27961-1GEA 1570 EUR
APOSPC27961-1G
1-(4-Fluorophenyl)-4-(4-methylpiperazin-1-yl)butan-1-one, tech
1-(4-Fluorophenyl)-4-(4-methylpiperazin-1-yl)butan-1-one
Formel: C₁₅H₂₁FN₂O
Molecular Weight: 264,338 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00826065
CAS-Nummer: 98897-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR