3-Amino-3-(2-fluorophenyl)propan-1-ol, tech

Lieferant: Apollo Scientific

PC27962-1G
APOSPC27962-1GEA 1570 EUR
APOSPC27962-1G
3-Amino-3-(2-fluorophenyl)propan-1-ol, tech
3-Amino-3-(2-fluorophenyl)propan-1-ol
Formel: C₉H₁₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD03412714
CAS-Nummer: 612532-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR