3-Amino-3-[3-(trifluoromethyl)phenyl]propan-1-ol, tech

Lieferant: Apollo Scientific

PC27963-1G
APOSPC27963-1GEA 0 EUR
APOSPC27963-1G
3-Amino-3-[3-(trifluoromethyl)phenyl]propan-1-ol, tech
3-Amino-3-[3-(trifluoromethyl)phenyl]propan-1-ol
Formel: C₁₀H₁₂F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03412708
CAS-Nummer: 683221-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR