(2E)-3-(Dimethylamino)-1-[3-fluoro-5-(trifluoromethyl)phenyl]prop-2-en-1-one, tech

Lieferant: Apollo Scientific

PC27964-1G
APOSPC27964-1GEA 1570 EUR
APOSPC27964-1G
(2E)-3-(Dimethylamino)-1-[3-fluoro-5-(trifluoromethyl)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[3-fluoro-5-(trifluoromethyl)phenyl]prop-2-en-1-one
Formel: C₁₂H₁₁F₄NO
Molecular Weight: 261,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496095
CAS-Nummer: 154258-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR