3-[(1E)-(Dimethylamino)methylidene]-1-(2-fluorophenyl)thiourea, tech
Lieferant: Apollo Scientific
PC27966-1G
APOSPC27966-1GEA
1570
EUR
APOSPC27966-1G
3-[(1E)-(Dimethylamino)methylidene]-1-(2-fluorophenyl)thiourea, tech
3-[(1E)-(Dimethylamino)methylidene]-1-(2-fluorophenyl)thiourea
Formel:
C₁₀H₁₂FN₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139327 CAS-Nummer: 477851-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...