3-[(1E)-(Dimethylamino)methylidene]-1-(4-fluorophenyl)thiourea, tech

Lieferant: Apollo Scientific

PC27967-1G
APOSPC27967-1GEA 1570 EUR
APOSPC27967-1G
3-[(1E)-(Dimethylamino)methylidene]-1-(4-fluorophenyl)thiourea, tech
3-[(1E)-(Dimethylamino)methylidene]-1-(4-fluorophenyl)thiourea
Formel: C₁₀H₁₂FN₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00139342
CAS-Nummer: 1661847-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR