1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile, tech

Lieferant: Apollo Scientific

PC27968-1G
APOSPC27968-1GEA 1570 EUR
APOSPC27968-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile, tech
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile
Formel: C₁₀H₄ClF₃N₄
Molecular Weight: 272,614 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664252
CAS-Nummer: 122890-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR