[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl 2-chloroacetate, tech

Lieferant: Apollo Scientific

PC27969-1G
APOSPC27969-1GEA 1570 EUR
APOSPC27969-1G
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl 2-chloroacetate, tech
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl 2-chloroacetate
Formel: C₈H₇Cl₂F₃N₂O₂
Molecular Weight: 291,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141485
CAS-Nummer: 303148-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR