[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl acetate, tech

Lieferant: Apollo Scientific

PC27970-1G
APOSPC27970-1GEA 1570 EUR
APOSPC27970-1G
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl acetate, tech
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl acetate
Formel: C₈H₈ClF₃N₂O₂
Molecular Weight: 256,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141486
CAS-Nummer: 303148-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR