3-{[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]oxy}butan-2-one, tech

Lieferant: Apollo Scientific

PC27972-1G
APOSPC27972-1GEA 1570 EUR
APOSPC27972-1G
3-{[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]oxy}butan-2-one, tech
3-{[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]oxy}butan-2-one
Formel: C₉H₁₁F₃N₂O₂
Molecular Weight: 236,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314996
CAS-Nummer: 477708-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR