N'-[(1E)-1-(4-Fluorophenyl)ethylidene]methoxycarbohydrazide, tech

Lieferant: Apollo Scientific

PC27973-1G
APOSPC27973-1GEA 1570 EUR
APOSPC27973-1G
N'-[(1E)-1-(4-Fluorophenyl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(4-Fluorophenyl)ethylidene]methoxycarbohydrazide
Formel: C₁₀H₁₁FN₂O₂
Molecular Weight: 210,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792964
CAS-Nummer: 112382-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR