N'-[(1E)-1-[3-(Trifluoromethyl)phenyl]ethylidene]methoxycarbohydrazide, tech
Lieferant: Apollo Scientific
PC27974-1G
APOSPC27974-1GEA
1570
EUR
APOSPC27974-1G
N'-[(1E)-1-[3-(Trifluoromethyl)phenyl]ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-[3-(Trifluoromethyl)phenyl]ethylidene]methoxycarbohydrazide
Formel:
C₁₁H₁₁F₃N₂O₂ Molecular Weight: 260,212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792965 CAS-Nummer: 303148-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...