1-(2-Fluorphenyl)pyrrol 95%

Lieferant: Apollo Scientific

PC27975-1G
APOSPC27975-1GEA 37.7 EUR
APOSPC27975-1G
1-(2-Fluorphenyl)pyrrol 95%
1-(2-Fluorophenyl)pyrrole
Formel: C₁₀H₈FN
Molecular Weight: 161,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02665231
CAS-Nummer: 89096-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR