(3Z)-1,1,1-Trifluoro-4-[(4-methoxyphenyl)amino]pent-3-en-2-one, tech

Lieferant: Apollo Scientific

PC27977-1G
APOSPC27977-1GEA 1570 EUR
APOSPC27977-1G
(3Z)-1,1,1-Trifluoro-4-[(4-methoxyphenyl)amino]pent-3-en-2-one, tech
(3Z)-1,1,1-Trifluoro-4-[(4-methoxyphenyl)amino]pent-3-en-2-one
Formel: C₁₂H₁₂F₃NO₂
Molecular Weight: 259,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568218
CAS-Nummer: 201659-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR