3-(Cyclopentylamino)-1,1,1-trifluoropropan-2-ol, tech

Lieferant: Apollo Scientific

PC27979-1G
APOSPC27979-1GEA 1570 EUR
APOSPC27979-1G
3-(Cyclopentylamino)-1,1,1-trifluoropropan-2-ol, tech
3-(Cyclopentylamino)-1,1,1-trifluoropropan-2-ol
Formel: C₈H₁₄F₃NO
Molecular Weight: 197,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082684
CAS-Nummer: 477858-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR