2,2,2-Trifluoro-N-[3-(trifluoromethyl)phenyl]acetamide 95%

Lieferant: Apollo Scientific

PC27980-1G
APOSPC27980-1GEA 874 EUR
APOSPC27980-1G
2,2,2-Trifluoro-N-[3-(trifluoromethyl)phenyl]acetamide 95%
2,2,2-Trifluoro-N-[3-(trifluoromethyl)phenyl]acetamide
Formel: C₉H₅F₆NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00444115
CAS-Nummer: 2946-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR