(3Z)-3-(1-{[2-(Trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one, tech

Lieferant: Apollo Scientific

PC27981-1G
APOSPC27981-1GEA 1570 EUR
APOSPC27981-1G
(3Z)-3-(1-{[2-(Trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one, tech
(3Z)-3-(1-{[2-(Trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one
Formel: C₁₃H₁₂F₃NO₂
Molecular Weight: 271,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617585
CAS-Nummer: 685108-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR