2-[4-(4-Fluorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Lieferant: Apollo Scientific
PC27982-1G
APOSPC27982-1GEA
1570
EUR
APOSPC27982-1G
2-[4-(4-Fluorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[4-(4-Fluorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel:
C₁₁H₉FN₄O Molecular Weight: 232,214 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787593 CAS-Nummer: 860651-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...