3-(3-Fluorophenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

PC27983-1G
APOSPC27983-1GEA 1570 EUR
APOSPC27983-1G
3-(3-Fluorophenoxy)butan-2-one, tech
3-(3-Fluorophenoxy)butan-2-one
Formel: C₁₀H₁₁FO₂
Molecular Weight: 182,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243584
CAS-Nummer: 306979-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR