3-(2,4-Difluorophenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

PC27984-1G
APOSPC27984-1GEA 1570 EUR
APOSPC27984-1G
3-(2,4-Difluorophenoxy)butan-2-one, tech
3-(2,4-Difluorophenoxy)butan-2-one
Formel: C₁₀H₁₀F₂O₂
Molecular Weight: 200,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243588
CAS-Nummer: 306979-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR