(1Z)-1-(Dimethylamino)-4-(3-fluorophenoxy)pent-1-en-3-one, tech

Lieferant: Apollo Scientific

PC27985-1G
APOSPC27985-1GEA 1570 EUR
APOSPC27985-1G
(1Z)-1-(Dimethylamino)-4-(3-fluorophenoxy)pent-1-en-3-one, tech
(1Z)-1-(Dimethylamino)-4-(3-fluorophenoxy)pent-1-en-3-one
Formel: C₁₃H₁₆FNO₂
Molecular Weight: 237,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243590
CAS-Nummer: 1164520-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR