(1E)-4-(2,4-Difluorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech

Lieferant: Apollo Scientific

PC27986-1G
APOSPC27986-1GEA 1570 EUR
APOSPC27986-1G
(1E)-4-(2,4-Difluorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech
(1E)-4-(2,4-Difluorophenoxy)-1-(dimethylamino)pent-1-en-3-one
Formel: C₁₃H₁₅F₂NO₂
Molecular Weight: 255,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243593
CAS-Nummer: 1164564-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR