3-(2-Fluorophenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

PC27987-1G
APOSPC27987-1GEA 1570 EUR
APOSPC27987-1G
3-(2-Fluorophenoxy)butan-2-one, tech
3-(2-Fluorophenoxy)butan-2-one
Formel: C₁₀H₁₁FO₂
Molecular Weight: 182,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243594
CAS-Nummer: 30343-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR