(3E)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech

Lieferant: Apollo Scientific

PC27988-1G
APOSPC27988-1GEA 1570 EUR
APOSPC27988-1G
(3E)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech
(3E)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one
Formel: C₁₅H₁₂FN₃O
Molecular Weight: 269.274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314820
CAS-Nummer: 320422-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR