3-[(2-Chloro-6-fluorophenyl)methyl]-1-cyclopropyl-4-hydroxy-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

PC27989-1G
APOSPC27989-1GEA 1570 EUR
APOSPC27989-1G
3-[(2-Chloro-6-fluorophenyl)methyl]-1-cyclopropyl-4-hydroxy-1,2-dihydropyridin-2-one, tech
3-[(2-Chloro-6-fluorophenyl)methyl]-1-cyclopropyl-4-hydroxy-1,2-dihydropyridin-2-one
Formel: C₁₅H₁₃ClFNO₂
Molecular Weight: 293,721 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871448
CAS-Nummer: 477860-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR