(Z)-(Ethyl N-[(Z)-4-(trifluoromethyl)benzoyl]ethanecarbohydrazonate), tech

Lieferant: Apollo Scientific

PC27990-1G
APOSPC27990-1GEA 1570 EUR
APOSPC27990-1G
(Z)-(Ethyl N-[(Z)-4-(trifluoromethyl)benzoyl]ethanecarbohydrazonate), tech
(Z)-(Ethyl N-[(Z)-4-(trifluoromethyl)benzoyl]ethanecarbohydrazonate)
Formel: C₁₂H₁₃F₃N₂O₂
Molecular Weight: 274,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082700
CAS-Nummer: 477862-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR