2-[4-(3-Fluoropropoxy)phenyl]acetonitrile, tech

Lieferant: Apollo Scientific

PC27991-1G
APOSPC27991-1GEA 1570 EUR
APOSPC27991-1G
2-[4-(3-Fluoropropoxy)phenyl]acetonitrile, tech
2-[4-(3-Fluoropropoxy)phenyl]acetonitrile
Formel: C₁₁H₁₂FNO
Molecular Weight: 193.217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082793
CAS-Nummer: 477862-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR