2-[4-(3-Fluorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech

Lieferant: Apollo Scientific

PC27992-1G
APOSPC27992-1GEA 1570 EUR
APOSPC27992-1G
2-[4-(3-Fluorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[4-(3-Fluorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel: C₁₁H₉FN₄O
Molecular Weight: 232,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787606
CAS-Nummer: 860786-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR