(3E)-1,1,1-Trifluoro-4-[(1-methylpiperidin-4-yl)amino]pent-3-en-2-one, tech

Lieferant: Apollo Scientific

PC27995-1G
APOSPC27995-1GEA 1570 EUR
APOSPC27995-1G
(3E)-1,1,1-Trifluoro-4-[(1-methylpiperidin-4-yl)amino]pent-3-en-2-one, tech
(3E)-1,1,1-Trifluoro-4-[(1-methylpiperidin-4-yl)amino]pent-3-en-2-one
Formel: C₁₁H₁₇F₃N₂O
Molecular Weight: 250.261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064709
CAS-Nummer: 939762-11-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR