(1E,4E)-1-(Dimethylamino)-5-(4-fluorophenyl)penta-1,4-dien-3-one, tech
Lieferant: Apollo Scientific
PC27996-1G
APOSPC27996-1GEA
1570
EUR
APOSPC27996-1G
(1E,4E)-1-(Dimethylamino)-5-(4-fluorophenyl)penta-1,4-dien-3-one, tech
(1E,4E)-1-(Dimethylamino)-5-(4-fluorophenyl)penta-1,4-dien-3-one
Formel:
C₁₃H₁₄FNO Molecular Weight: 219,255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172120 CAS-Nummer: 320416-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...