6-[(2-Fluorophenyl)methyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

PC27997-1G
APOSPC27997-1GEA 1570 EUR
APOSPC27997-1G
6-[(2-Fluorophenyl)methyl]pyridazin-3-ol, tech
6-[(2-Fluorophenyl)methyl]pyridazin-3-ol
Formel: C₁₁H₉FN₂O
Molecular Weight: 204,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202114
CAS-Nummer: 200001-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR