2-(Morpholin-4-yl)-2-[3-(trifluoromethyl)phenyl]acetonitrile, tech

Lieferant: Apollo Scientific

PC27999-1G
APOSPC27999-1GEA 1570 EUR
APOSPC27999-1G
2-(Morpholin-4-yl)-2-[3-(trifluoromethyl)phenyl]acetonitrile, tech
2-(Morpholin-4-yl)-2-[3-(trifluoromethyl)phenyl]acetonitrile
Formel: C₁₃H₁₃F₃N₂O
Molecular Weight: 270,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173086
CAS-Nummer: 66548-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR