2-[(2-Fluorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one, tech
Lieferant: Apollo Scientific
PC28010-1G
APOSPC28010-1GEA
1570
EUR
APOSPC28010-1G
2-[(2-Fluorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one, tech
2-[(2-Fluorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one
Formel:
C₁₄H₁₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD02083019 CAS-Nummer: 477871-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...